Merck
  • Intermolecular methoxycarbonylation of terminal alkynes catalyzed by palladium(II) bis(oxazoline) complexes.
Intermolecular methoxycarbonylation of terminal alkynes catalyzed by palladium(II) bis(oxazoline) complexes.
Angewandte Chemie (International ed. in English) (2009-02-05)
Keisuke Kato, Satoshi Motodate, Tomoyuki Mochida, Takuya Kobayashi, Hiroyuki Akita
ABSTRACT
Boxing clever: Direct conversion of a terminal alkyne group into a beta-methoxyacrylate is realized with the aid of the bis(oxazoline) ligand (box). Acetyl and ketal protecting groups, free hydroxy groups, and acid-sensitive glycosidic bonds are not affected under the reaction conditions. The one-pot synthesis of (+/-)-dihydrokawain from the homopropargyl alcohol is also achieved. tfa = trifluoroacetate.
Sigma-Aldrich
Palladium(II) trifluoroacetate, 97%