Merck
  • Kinetic Studies on Formic Acid Dehydrogenation Catalyzed by an Iridium Complex towards Insights into the Catalytic Mechanism of High-Pressure Hydrogen Gas Production.
Kinetic Studies on Formic Acid Dehydrogenation Catalyzed by an Iridium Complex towards Insights into the Catalytic Mechanism of High-Pressure Hydrogen Gas Production.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-05)
Masayuki Iguchi, Heng Zhong, Yuichiro Himeda, Hajime Kawanami
ABSTRACT
Kinetic studies on the catalytic reaction mechanism of formic acid (FA) dehydrogenation were performed in the presence of a water-soluble iridium complex bearing a 4,4'-dihydroxy-2,2'-bipyridine ligand. Determination of kinetic isotope effects revealed that a shift of the rate-limiting step at low and high concentrations of FA can be caused by the pH dependence of the reaction steps. The proposed equation for the reaction rate corresponds well with the experimental results concerning the shift phenomena. Towards industrial application in future hydrogen fueling stations, this will able the design of a dehydrogenation system catalyzed by the iridium complex.