Merck
  • The Multiple Localized Glyceraldehyde-3-Phosphate Dehydrogenase Contributes to the Attenuation of the
The Multiple Localized Glyceraldehyde-3-Phosphate Dehydrogenase Contributes to the Attenuation of the
Frontiers in cellular and infection microbiology (2018-01-13)
Ivona Pavkova, Monika Kopeckova, Jana Klimentova, Monika Schmidt, Valeria Sheshko, Margarita Sobol, Jitka Zakova, Pavel Hozak, Jiri Stulik
ABSTRACT
The DsbA homolog of
Sigma-Aldrich
D-Ala-D-Ala