Merck
  • Ultrafast all-climate aluminum-graphene battery with quarter-million cycle life.
Ultrafast all-climate aluminum-graphene battery with quarter-million cycle life.
Science advances (2017-12-20)
Hao Chen, Hanyan Xu, Siyao Wang, Tieqi Huang, Jiabin Xi, Shengying Cai, Fan Guo, Zhen Xu, Weiwei Gao, Chao Gao
ABSTRACT
Rechargeable aluminum-ion batteries are promising in high-power density but still face critical challenges of limited lifetime, rate capability, and cathodic capacity. We design a "trihigh tricontinuous" (3H3C) graphene film cathode with features of high quality, orientation, and channeling for local structures (3H) and continuous electron-conducting matrix, ion-diffusion highway, and electroactive mass for the whole electrode (3C). Such a cathode retains high specific capacity of around 120 mAh g
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)