Merck
  • Fabrication of charge reversible graphene oxide-based nanocomposite with multiple antibacterial modes and magnetic recyclability.
Fabrication of charge reversible graphene oxide-based nanocomposite with multiple antibacterial modes and magnetic recyclability.
Journal of colloid and interface science (2017-10-17)
Qiang Li, Chunyan Yong, Weiwei Cao, Xi Wang, Lina Wang, Jie Zhou, Xiaodong Xing
ABSTRACT
In this work, N-alkylated poly (4-vinylpyridine) (NPVP), a cationic polymer, was firstly applied for the surface modification of Fe
Sigma-Aldrich
(3-Bromopropyl)trimethoxysilane, ≥97.0%