Merck
In vitro modeling of HIV proviral activity in microglia.
The FEBS journal (2017-11-09)
Lee A Campbell, Christopher T Richie, Yajun Zhang, Emily J Heathward, Lamarque M Coke, Emily Y Park, Brandon K Harvey
ABSTRACT
Microglia, the resident macrophages of the brain, play a key role in the pathogenesis of HIV-associated neurocognitive disorders (HAND) due to their productive infection by HIV. This results in the release of neurotoxic viral proteins and pro-inflammatory compounds which negatively affect the functionality of surrounding neurons. Because models of HIV infection within the brain are limited, we aimed to create a novel microglia cell line with an integrated HIV provirus capable of recreating several hallmarks of HIV infection. We utilized clustered regularly interspaced short palindromic repeats (CRISPR)/Cas9 gene editing technology and integrated a modified HIV provirus into CHME-5 immortalized microglia to create HIV-NanoLuc CHME-5. In the modified provirus, the Gag-Pol region is replaced with the coding region for NanoLuciferase (NanoLuc), which allows for the rapid assay of HIV long terminal repeat activity using a luminescent substrate, while still containing the necessary genetic material to produce established neurotoxic viral proteins (e.g. tat, nef, gp120). We confirmed that HIV-NanoLuc CHME-5 microglia express NanoLuc, along with the HIV viral protein Nef. We subsequently exposed these cells to a battery of experiments to modulate the activity of the provirus. Proviral activity was enhanced by treating the cells with pro-inflammatory factors lipopolysaccharide (LPS) and tumor necrosis factor alpha and by overexpressing the viral regulatory protein Tat. Conversely, genetic modification of the toll-like receptor-4 gene by CRISPR/Cas9 reduced LPS-mediated proviral activation, and pharmacological application of NF-κB inhibitor sulfasalazine similarly diminished proviral activity. Overall, these data suggest that HIV-NanoLuc CHME-5 may be a useful tool in the study of HIV-mediated neuropathology and proviral regulation.