Merck
  • Hepatic glucuronidation of 4-tert-octylphenol in humans: inter-individual variability and responsible UDP-glucuronosyltransferase isoforms.
Hepatic glucuronidation of 4-tert-octylphenol in humans: inter-individual variability and responsible UDP-glucuronosyltransferase isoforms.
Archives of toxicology (2017-05-14)
Takashi Isobe, Susumu Ohkawara, Toshiko Tanaka-Kagawa, Hideto Jinno, Nobumitsu Hanioka
ABSTRACT
4-tert-Octylphenol (4-tOP) is an endocrine-disrupting chemical. It is mainly metabolized into glucuronide by UDP-glucuronosyltransferase (UGT) enzymes in humans. The purpose of this study was to assess inter-individual variability in and the possible roles of UGT isoforms in hepatic 4-tOP glucuronidation in the humans. 4-tOP glucuronidation activities in the liver microsomes and recombinant UGTs of humans were assessed at broad substrate concentrations, and kinetics were analyzed. Correlation analyses between 4-tOP and diclofenac or 4-hydroxybiphenyl activities in pooled and individual human liver microsomes were also performed. Typical CL
Sigma-Aldrich
4-Pentylphenol, ≥98.0% (GC)