Merck
  • The influence of seawater properties on toxicity of copper pyrithione and its degradation product to brine shrimp Artemia salina.
The influence of seawater properties on toxicity of copper pyrithione and its degradation product to brine shrimp Artemia salina.
Ecotoxicology and environmental safety (2017-08-26)
Vesna Lavtizar, Daisuke Kimura, Satoshi Asaoka, Hideo Okamura
ABSTRACT
Copper pyrithione (CuPT) is a biocide, used worldwide to prevent biofouling on submerged surfaces. In aquatic environments it rapidly degrades, however, one of the degradation products (HPT) is known to react with cupric ion back to its parent compound. Not much is known about the behavior and toxicity of CuPT and its degradation product HPT in different water systems. Hence, our aim was to investigate the ecotoxicity of CuPT, HPT as well as Cu
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%