Merck
  • Overexpression, purification and enzymatic characterization of a recombinant Arabian camel Camelus dromedarius glucose-6-phosphate dehydrogenase.
Overexpression, purification and enzymatic characterization of a recombinant Arabian camel Camelus dromedarius glucose-6-phosphate dehydrogenase.
Protein expression and purification (2015-09-13)
Hesham Saeed, Mohammad Ismaeil, Amira Embaby, Farid Ataya, Ajamaluddin Malik, Manal Shalaby, Sabah El-Banna, Ahmed Abdelrahim Mohamed Ali, Khalid Bassiouny
ABSTRACT
In a previous study the full-length open reading frame of the Arabian camel, Camelus dromedarius liver cytosolic glucose-6-phosphate dehydrogenase (G6PD) cDNA was determined using reverse transcription polymerase chain reaction. The C. dromedarius cDNA was found to be 1545 nucleotides (accession number JN098421) that encodes a protein of 515 amino acids residues. In the present study, C. dromedarius recombinant G6PD was heterologously overexpressed in Escherichia coli BL21 (DE3) pLysS and purified by immobilized metal affinity fast protein liquid chromatography (FPLC) in a single step. The purity and molecular weight of the enzyme were analyzed on SDS-PAGE and the purified enzyme showed a single band on the gel with a molecular weight of 63.0 KDa. The specific activity was determined to be 2000 EU/mg protein. The optimum temperature and pH were found to be 60 °C and 7.4, respectively. The isoelectric point (pI) for the purified G6PD was determined to be 6.4. The apparent K