Merck
Primary cortical neurons on PMCS TiO
Biophysical chemistry (2017-05-10)
Alessandro Roncador, Aura Matilde Jimenez-Garduño, Laura Pasquardini, Giovanni Giusti, Nicola Cornella, Lorenzo Lunelli, Cristina Potrich, Ruben Bartali, Lucrezia Aversa, Roberto Verucchi, Mauro Dalla Serra, Silvia Caponi, Salvatore Iannotta, Paolo Macchi, Carlo Musio
ABSTRACT
We report a comprehensive study of the biocompatibility and neurocompatibility of titanium dioxide films (TiO
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline