Merck
  • A small-scale, portable method for extracting microplastics from marine sediments.
A small-scale, portable method for extracting microplastics from marine sediments.
Environmental pollution (Barking, Essex : 1987) (2017-07-25)
Rachel L Coppock, Matthew Cole, Penelope K Lindeque, Ana M Queirós, Tamara S Galloway
ABSTRACT
Microplastics (plastic particles, 0.1 μm-5 mm in size) are widespread marine pollutants, accumulating in benthic sediments and shorelines the world over. To gain a clearer understanding of microplastic availability to marine life, and the risks they pose to the health of benthic communities, ecological processes and food security, it is important to obtain accurate measures of microplastic abundance in marine sediments. To date, methods for extracting microplastics from marine sediments have been disadvantaged by complexity, expense, low extraction efficiencies and incompatibility with very fine sediments. Here we present a new, portable method to separate microplastics from sediments of differing types, using the principle of density floatation. The Sediment-Microplastic Isolation (SMI) unit is a custom-built apparatus which consistently extracted microplastics from sediments in a single step, with a mean efficiency of 95.8% (±SE 1.6%; min 70%, max 100%). Zinc chloride, at a density of 1.5 g cm
Product Description
Zipper-top poly bag, size 5 in. × 8 in.