Merck
  • In vitro toxicity of perfluorooctane sulfonate on rat liver hepatocytes: probability of distructive binding to CYP 2E1 and involvement of cellular proteolysis.
In vitro toxicity of perfluorooctane sulfonate on rat liver hepatocytes: probability of distructive binding to CYP 2E1 and involvement of cellular proteolysis.
Environmental science and pollution research international (2017-08-27)
Mehdi Rajabnia Khansari, Bahareh Sadat Yousefsani, Farzad Kobarfard, Mehrdad Faizi, Jalal Pourahmad
ABSTRACT
Perfluorooctanesulfonate (PFOS), an anthropogenic fluorosurfactant, is one of the most common global pollutants. PFOS is used in various consumer products to provide soil, oil, and water resistance to materials used in clothing, upholstery, and food packaging. PFOS is persistent, bioaccumulative, and toxic to mammalian species. In this study, the cellular mechanisms involved in PFOS hepatotoxicity were evaluated. For this purpose, we determined oxidative stress markers including cell lysis, ROS generation, lipid peroxidation, glutathione depletion, mitochondrial membrane potential decrease, lysosomal membrane leakiness, and cellular proteolysis. Our results demonstrated that PFOS liver cytotoxicity was associated with reactive oxygen species (ROS) formation and lipid peroxidation in isolated rat hepatocytes. Incubation of hepatocytes with PFOS caused rapid depletion of hepatocyte glutathione (GSH), an important marker of cellular oxidative stress. Most of the PFOS-induced GSH depletion could be attributed to the expulsion of glutathione disulfide (GSSG). PFOS hepatotoxicity was inhibited by antioxidants and ROS scavengers, mitochondrial permeability transition (MPT) pore sealing agents, and endocytosis inhibitors. Our results suggest that PFOS hepatotoxicity might be the result of oxidative stress-induced lysosomal membrane leakiness and cellular proteolysis in rat hepatocytes.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Bromoheptane, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Phenylimidazole, 97%
Pricing and availability is not currently available.