Merck
  • Whole-body propionate and glucose metabolism of multiparous dairy cows receiving folic acid and vitamin B
Whole-body propionate and glucose metabolism of multiparous dairy cows receiving folic acid and vitamin B
Journal of dairy science (2017-08-07)
M Duplessis, H Lapierre, B Ouattara, N Bissonnette, D Pellerin, J-P Laforest, C L Girard
ABSTRACT
This study was undertaken to evaluate the effect of supplementation of folic acid and vitamin B
Sigma-Aldrich
5-Sulfosalicylic acid hydrate, 95%