Merck
  • Enhanced Second-Harmonic Generation from Sequential Capillarity-Assisted Particle Assembly of Hybrid Nanodimers.
Enhanced Second-Harmonic Generation from Sequential Capillarity-Assisted Particle Assembly of Hybrid Nanodimers.
Nano letters (2017-08-03)
Flavia Timpu, Nicholas R Hendricks, Mihail Petrov, Songbo Ni, Claude Renaut, Heiko Wolf, Lucio Isa, Yuri Kivshar, Rachel Grange
ABSTRACT
We show enhanced second-harmonic generation (SHG) from a hybrid metal-dielectric nanodimer consisting of an inorganic perovskite nanoparticle of barium titanate (BaTiO
Sigma-Aldrich
Chlorodimethylsilane, 98%
Sigma-Aldrich
Triton X-45