Merck
  • Gankyrin induces STAT3 activation in tumor microenvironment and sorafenib resistance in hepatocellular carcinoma.
Gankyrin induces STAT3 activation in tumor microenvironment and sorafenib resistance in hepatocellular carcinoma.
Cancer science (2017-08-05)
Toshiharu Sakurai, Norihisa Yada, Satoru Hagiwara, Tadaaki Arizumi, Kosuke Minaga, Ken Kamata, Mamoru Takenaka, Yasunori Minami, Tomohiro Watanabe, Naoshi Nishida, Masatoshi Kudo
ABSTRACT
Most hepatocellular carcinomas (HCC) develop as a result of chronic liver inflammation. We have shown that the oncoprotein gankyrin is critical for inflammation-induced tumorigenesis in the colon. Although the in vitro function of gankyrin is well known, its role in vivo remains to be elucidated. We investigated the effect of gankyrin in the tumor microenvironment of mice with liver parenchymal cell-specific gankyrin ablation (Alb-Cre;gankyrin
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMD10