- The Stimulation of Macrophages with TLR Ligands Supports Increased IL-19 Expression in Inflammatory Bowel Disease Patients and in Colitis Models.
The Stimulation of Macrophages with TLR Ligands Supports Increased IL-19 Expression in Inflammatory Bowel Disease Patients and in Colitis Models.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-03)
Anna Steinert, Ioannis Linas, Berna Kaya, Mohamed Ibrahim, Andreas Schlitzer, Petr Hruz, Katarina Radulovic, Luigi Terracciano, Andrew J Macpherson, Jan Hendrik Niess
PMID28864472
ABSTRACT
IL-19, a member of the IL-10 cytokine family that signals through the IL-20 receptor type I (IL-20Rα:IL-20Rβ), is a cytokine whose function is not completely known. In this article, we show that the expression of
MATERIALS