Merck
H
Oncotarget (2017-10-17)
Zhiwen Zeng, Dejun Wang, Uma Gaur, Liao Rifang, Haitao Wang, Wenhua Zheng
ABSTRACT
Impairment of insulin-like growth factor I (IGF-I) signaling plays an important role in the development of neurodegeneration. In the present study, we investigated the effect of H
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
CPCCOEt, ≥98% (HPLC)