Merck
  • Regulation of calcineurin activity in insulin-secreting cells: stimulation by Hsp90 during glucocorticoid-induced apoptosis.
Regulation of calcineurin activity in insulin-secreting cells: stimulation by Hsp90 during glucocorticoid-induced apoptosis.
Cellular signalling (2008-07-10)
Felicia Ranta, Martina Düfer, Björn Stork, Sebastian Wesselborg, Gisela Drews, Hans-Ulrich Häring, Florian Lang, Susanne Ullrich
ABSTRACT
Previously, we described that apoptotic cell death induced by the synthetic glucocorticoid dexamethasone (dex) is inhibited by calcineurin inhibitors, FK506 and deltamethrin, in insulin-secreting cells. The aim of the present study was to examine the mechanism of dex-dependent activation of calcineurin. In INS-1 cells cultured up to 4d with dex (100 nmol/l), the percentage of apoptosis, quantified by condensed nuclei and TUNEL positive cells, increased from 1% to 10.9%. FK506 inhibited dex-mediated cell death. Apoptosis was significantly higher at glucose concentrations that induce [Ca(2+)](i) oscillations than at low, non-stimulatory glucose. Dex had no acute effect on [Ca(2+)](i). Calcineurin activity, measured in control and dex-treated cell homogenates, revealed that maximal activity and the sensitivity to the substrate RII peptide was unaltered. However, dex treatment significantly increased enzyme activity at submaximal, physiological Ca(2+) concentrations. Dex did not stimulate the Ca(2+)-dependent protease calpain, known to activate calcineurin by cleavage, as no cleaved calcineurin was detectable. Furthermore, the calpain inhibitor ALLN did not counteract dex-dependent cell death. Western blotting revealed that in dex-treated cells heat shock protein 90 (Hsp90), a component of the glucocorticoid receptor (GR) known to stimulate calcineurin, was increased while calcineurin protein levels were unchanged. In immunoprecipitates with calcineurin antibodies, Hsp90 was only detected in dex-treated cell homogenates. These data suggest that dex-induced apoptosis involves release of Hsp90 from the stimulated GR complex, subsequent binding to and activation of calcineurin, that may contribute to dex-mediated cell death in the presence of high glucose.