- Inflammasome and Fas-Mediated IL-1β Contributes to Th17/Th1 Cell Induction in Pathogenic Bacterial Infection In Vivo.
Inflammasome and Fas-Mediated IL-1β Contributes to Th17/Th1 Cell Induction in Pathogenic Bacterial Infection In Vivo.
Journal of immunology (Baltimore, Md. : 1950) (2017-07-05)
Ryosuke Uchiyama, Shin Yonehara, Shun'ichiro Taniguchi, Satoshi Ishido, Ken J Ishii, Hiroko Tsutsui
PMID28674179
ABSTRACT
CD4