Merck
Ethnopharmacological and Chemical Characterization of
Frontiers in pharmacology (2017-08-10)
Vanessa Martínez-Francés, Emeline Hahn, Segundo Ríos, Diego Rivera, Eike Reich, Roser Vila, Salvador Cañigueral
ABSTRACT
In Valencia Region (Spain), some wild and cultivated sages are used for medicinal purposes. Among them,
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 400
Pricing and availability is not currently available.