Merck
  • ATP2A3 gene as an important player for resveratrol anticancer activity in breast cancer cells.
ATP2A3 gene as an important player for resveratrol anticancer activity in breast cancer cells.
Molecular carcinogenesis (2017-02-06)
Eduardo Izquierdo-Torres, Gabriela Rodríguez, Iván Meneses-Morales, Angel Zarain-Herzberg
ABSTRACT
The Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP2A2
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP2A3