Merck
Compartmentalized cAMP responses to prostaglandin EP
British journal of pharmacology (2017-06-13)
Shailesh R Agarwal, Kathryn Miyashiro, Htun Latt, Rennolds S Ostrom, Robert D Harvey
ABSTRACT
Previous studies indicate that prostaglandin EP FRET-based cAMP biosensors were targeted to different subcellular locations of primary human ASM cells. The Epac2-camps biosensor, which expresses throughout the cell, was used to measure bulk cytoplasmic responses. Epac2-MyrPalm and Epac2-CAAX were used to measure responses associated with lipid raft and non-raft regions of the plasma membrane respectively. Epac2-NLS was used to monitor responses at the nucleus. Activation of AC with forskolin or β EP
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)