Merck
  • Inhibition of β-Glucocerebrosidase Activity Preserves Motor Unit Integrity in a Mouse Model of Amyotrophic Lateral Sclerosis.
Inhibition of β-Glucocerebrosidase Activity Preserves Motor Unit Integrity in a Mouse Model of Amyotrophic Lateral Sclerosis.
Scientific reports (2017-07-14)
Alexandre Henriques, Mylene Huebecker, Hélène Blasco, Céline Keime, Christian R Andres, Philippe Corcia, David A Priestman, Frances M Platt, Michael Spedding, Jean-Philippe Loeffler
ABSTRACT
Recent metabolomic reports connect dysregulation of glycosphingolipids, particularly ceramide and glucosylceramide, to neurodegeneration and to motor unit dismantling in amyotrophic lateral sclerosis at late disease stage. We report here altered levels of gangliosides in the cerebrospinal fluid of amyotrophic lateral sclerosis patients in early disease stage. Conduritol B epoxide is an inhibitor of acid beta-glucosidase, and lowers glucosylceramide degradation. Glucosylceramide is the precursor for all of the more complex glycosphingolipids. In SOD1
Sigma-Aldrich
β-Glucosidase Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
CTB, ≥98% (HPLC)