- CXCR5
CXCR5
Experimental cell research (2017-04-17)
Junjie Xing, Chenxin Zhang, Xiaohong Yang, Shaoxuan Wang, Zhongchuan Wang, Xu Li, Enda Yu
PMID28412245
ABSTRACT
Colorectal cancer is the third most prevalent cancer type worldwide and contributes to a significant percentage of cancer-related mortality. Recent studies have shown that the CXCR5
MATERIALS