Merck
  • DNA double-strand breaks and Aurora B mislocalization induced by exposure of early mitotic cells to H
DNA double-strand breaks and Aurora B mislocalization induced by exposure of early mitotic cells to H
Free radical biology & medicine (2017-03-28)
Min-Guk Cho, Ju-Hyun Ahn, Hee-Song Choi, Jae-Ho Lee
ABSTRACT
Aneuploidy, an abnormal number of chromosomes that is a hallmark of cancer cells, can arise from tetraploid/binucleated cells through a failure of cytokinesis. Reactive oxygen species (ROS) have been implicated in various diseases, including cancer. However, the nature and role of ROS in cytokinesis progression and related mechanisms has not been clearly elucidated. Here, using time-lapse analysis of asynchronously growing cells and immunocytochemical analyses of synchronized cells, we found that hydrogen peroxide (H
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture