Merck
  • Combined treatment with Metformin and 2-deoxy glucose induces detachment of viable MDA-MB-231 breast cancer cells in vitro.
Combined treatment with Metformin and 2-deoxy glucose induces detachment of viable MDA-MB-231 breast cancer cells in vitro.
Scientific reports (2017-05-13)
Maruša Bizjak, Petra Malavašič, Klemen Dolinar, Jelka Pohar, Sergej Pirkmajer, Mojca Pavlin
ABSTRACT
Triple naegative breast cancer has an increased rate of distant metastasis and consequently poor prognosis. To metastasize, breast cancer cells must detach from the main tumour mass and resist anoikis, a programmed cell death induced by lack of cell-extracellular matrix communication. Although cancer cells must detach to metastasize in vivo, the viability of floating cancer cells in vitro is rarely investigated. Here we show that co-treatment of anoikis-resistant MDA-MB-231 cells with metformin and 2-deoxy-D-glucose (2-DG) increased the percentage of floating cells, of which about 95% were viable. Floating cells resumed their proliferation once they were reseeded in the pharmacological compound-free medium. Similar effects on detachment were observed on anoikis-prone MCF-7 cells. Co-treatment of MDA-MB-231 cells with metformin and 2-DG induced a strong activation of AMP-activated protein kinase (AMPK), which was reduced by AMPK inhibitor compound C that prevented detachment of MDA-MB-231 cells. However, direct AMPK activators A-769662 and AICAR did not have any major effect on the percentage of floating MDA-MB-231 cells, indicating that AMPK activation is necessary but not sufficient for triggering detachment of cancer cells. Our results demonstrate that separate analysis of floating and attached cancer cells might be important for evaluation of anti-cancer agents.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture