Merck
  • Target analysis of tert-butyldimethylsilyl derivatives of nerve agent hydrolysis products by selectable one-dimensional or two-dimensional gas chromatography-mass spectrometry.
Target analysis of tert-butyldimethylsilyl derivatives of nerve agent hydrolysis products by selectable one-dimensional or two-dimensional gas chromatography-mass spectrometry.
Journal of chromatography. A (2017-04-25)
Yasuo Seto, Masumi Tachikawa, Mieko Kanamori-Kataoka, Kikuo Sasamoto, Nobuo Ochiai
ABSTRACT
A target analysis method for the sensitive and discriminative determination of the nerve agent hydrolysis products alkyl methylphosphonic acids as their tert-butyldimethylsilyl (TBDMS) derivatives was developed using a combination of selectable one- and two-dimensional (
Sigma-Aldrich
Methylphosphonic dichloride, 98%