Merck
  • Arrestins contribute to amyloid beta-induced cell death via modulation of autophagy and the α7nAch receptor in SH-SY5Y cells.
Arrestins contribute to amyloid beta-induced cell death via modulation of autophagy and the α7nAch receptor in SH-SY5Y cells.
Scientific reports (2017-06-15)
Yi-Qing Liu, Meng-Qi Jia, Zhao-Hong Xie, Xiao-Fei Liu, Hui-Yang, Xiao-Lei Zheng, Hui-Qing Yuan, Jian-Zhong Bi
ABSTRACT
Amyloid β-protein (Aβ) is believed to contribute to the development of Alzheimer's disease (AD). Here we showed that Aβ
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human CHRFAM7A