Merck
  • Cytotoxicity and genotoxicity effects of arsenic trioxide on SQ20B human laryngeal carcinoma cells.
Cytotoxicity and genotoxicity effects of arsenic trioxide on SQ20B human laryngeal carcinoma cells.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2017-03-07)
Fatma Trabelsi, Rim Khlifi, Didier Goux, Marilyne Guillamin, Amel Hamza-Chaffai, François Sichel
ABSTRACT
This study investigates the cytotoxicity and the genotoxicity induced by arsenic trioxide As
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass