Merck
  • Specific humoral and cellular immune responses in cancer patients undergoing chronic immunization with a VEGF-based therapeutic vaccine.
Specific humoral and cellular immune responses in cancer patients undergoing chronic immunization with a VEGF-based therapeutic vaccine.
Vaccine (2017-05-26)
Yanelys Morera, Javier Sánchez, Mónica Bequet-Romero, Katty-Hind Selman-Housein, Ana de la Torre, Francisco Hernández-Bernal, Yenima Martín, Acralys Garabito, Jesús Piñero, Cimara Bermúdez, Josué de la Torre, Marta Ayala, Jorge V Gavilondo
ABSTRACT
CIGB-247 is a cancer therapeutic vaccine, based on recombinant modified human vascular endothelial growth factor (VEGF) as antigen, in combination with the adjuvant VSSP, a bacterially-derived adjuvant. The vaccine have demonstrated efficacy in several murine malignancy models. These studies supported the rationale for a phase I clinical trial where safety, tolerance, and immunogenicity of CIGB-247 was studied in patients with advanced solid tumors at three antigen dose level. Surviving individuals of this clinical trial were eligible to receive off-trial voluntary re-immunizations. The present work is focus in the immunological follow up of these patients after approximately three years of immunizations, without additional oncological treatments. Long term vaccination was feasible and safe. Our results indicated that after sustained vaccination most of the patients conserved their seroconversion status. The specific anti-VEGF IgG titer diminished, but in all the cases keeps values up from the pre-vaccination levels. Continued vaccination was also important to produce a gradual shift in the anti-VEGF IgG response from IgG1 to Ig4. Outstanding, our results indicated that long-term off-trial vaccination could be associated with the maintaining of one reserve of antibodies able to interfere with the VEGF/Receptor interaction and the production of IFNγ secretion in CD8
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder