Merck
  • Selective B(4)-H Activation of an o-Carboranylthioamide Based on a Palladium Precursor.
Selective B(4)-H Activation of an o-Carboranylthioamide Based on a Palladium Precursor.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-11-30)
Yin-Ping Wang, Long Zhang, Yue-Jian Lin, Zhen-Hua Li, Guo-Xin Jin
ABSTRACT
Palladium(II)-induced selective B(4)-H activation of an o-carboranylthioamide has been developed. A tetranuclear palladium(II) complex has been obtained in high yield with excellent regioselectivity. DFT calculations have confirmed that the B(4)-borometalate is lower in energy than the corresponding B(3)-borometalate. The product proved to be a good precursor for target mononuclear or trinuclear B(4)-H activated complexes.