Merck
  • DHA-derived oxylipins, neuroprostanes and protectins, differentially and dose-dependently modulate the inflammatory response in human macrophages: Putative mechanisms through PPAR activation.
DHA-derived oxylipins, neuroprostanes and protectins, differentially and dose-dependently modulate the inflammatory response in human macrophages: Putative mechanisms through PPAR activation.
Free radical biology & medicine (2016-12-19)
Rémy Bosviel, Laurie Joumard-Cubizolles, Giulia Chinetti-Gbaguidi, Dominique Bayle, Corinne Copin, Nathalie Hennuyer, Isabelle Duplan, Bart Staels, Giuseppe Zanoni, Alessio Porta, Laurence Balas, Jean-Marie Galano, Camille Oger, Andrzej Mazur, Thierry Durand, Cécile Gladine
ABSTRACT
Whereas the anti-inflammatory properties and mechanisms of action of long chain ω3 PUFAs have been abundantly investigated, research gaps remain regarding the respective contribution and mechanisms of action of their oxygenated metabolites collectively known as oxylipins. We conducted a dose-dependent and comparative study in human primary macrophages aiming to compare the anti-inflammatory activity of two types of DHA-derived oxylipins including the well-described protectins (NPD1 and PDX), formed through lipoxygenase pathway and the neuroprostanes (14-A
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)