Merck
  • Metal Complexation Properties of Schiff Bases Containing 1,3,5-Triazine Derived from 2-Hydroxy-1-Naphthaldehyde in Solution. A Simple Spectrofluorimetric Method to Determine Mercury (II).
Metal Complexation Properties of Schiff Bases Containing 1,3,5-Triazine Derived from 2-Hydroxy-1-Naphthaldehyde in Solution. A Simple Spectrofluorimetric Method to Determine Mercury (II).
Journal of fluorescence (2016-09-17)
Miraç Ocak, Tuğba Ak, Ayşe Aktaş, Nurhayat Özbek, Osman Can Çağılcı, Abidin Gümrükçüoğlu, Halit Kantekin, Ümmühan Ocak, Hakan Alp
ABSTRACT
Four new Schiff base ligands carrying naphthalene groups were prepared from the reaction of 2,4-diamino-6-methyl-1,3,5-triazine and 2,4-diamino-6-undecyl-1,3,5-triazine with 2-hydroxy-1-naphthaldehyde. The influence of a series of metal ions including Cu
Sigma-Aldrich
1-Naphthaldehyde, 95%