Merck
VCP/p97 regulates β
Cellular signalling (2016-11-27)
Richard Wargachuk, Irina Glazkova, Nicolas Audet, Darlaine Pétrin, Phan Trieu, Terence E Hébert
ABSTRACT
GPCRs form signalling complexes with other receptors as part of dimers, G proteins and effector partners. A proteomic screen to identify proteins that associate with the β
Sigma-Aldrich
MISSION® esiRNA, targeting human VCP
Pricing and availability is not currently available.
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Pricing and availability is not currently available.
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Pricing and availability is not currently available.