Merck
  • Polyuria-associated hydronephrosis induced by xenobiotic chemical exposure in mice.
Polyuria-associated hydronephrosis induced by xenobiotic chemical exposure in mice.
American journal of physiology. Renal physiology (2016-07-22)
Wataru Yoshioka, Tatsuya Kawaguchi, Noriko Nishimura, Toshiya Akagi, Nozomi Fujisawa, Hiroyuki Yanagisawa, Fumio Matsumura, Chiharu Tohyama
ABSTRACT
Hydronephrosis is a commonly found disease state characterized by the dilation of renal calices and pelvis, resulting in the loss of kidney function in the severest cases. A generally accepted etiology of hydronephrosis involves the obstruction of urine flow along the urinary tract. In the recent years, we have developed a mouse model of hydronephrosis induced by lactational exposure to dioxin and demonstrated a lack of anatomical obstruction in this model. We also showed that prostaglandin E2 synthesis system plays a critical role in the onset of hydronephrosis. In the present study, we found that neonatal hydronephrosis was not likely to be associated with functional obstruction (impaired peristalsis) but was found to be associated with polyuria and low urine osmolality with the downregulation of proteins involved in the urine concentrating process. The administration of an antidiuretic hormone analog to the dioxin-exposed pups not only suppressed the increased urine output but also decreased the incidence and severity of hydronephrosis. In contrast to the case in pups, administration of dioxin to adult mice failed to induce polyuria and upregulation of prostaglandin E2 synthesis system, and the adult mice were resistant to develop hydronephrosis. These findings suggest the possibility that polyuria could induce hydronephrosis in the absence of anatomical or functional obstruction of the ureter. It is concluded that the present animal model provides a unique example of polyuria-associated type of hydronephrosis, suggesting a need to redefine this disease state.
Sigma-Aldrich
[deamino-Cys1, D-Arg8]-Vasopressin acetate salt hydrate, ≥95% (HPLC)