- Indirect Solid Freeform Fabrication of an Initiator-Free Photocrosslinkable Hydrogel Precursor for the Creation of Porous Scaffolds.
Indirect Solid Freeform Fabrication of an Initiator-Free Photocrosslinkable Hydrogel Precursor for the Creation of Porous Scaffolds.
In the present work, a photopolymerized urethane-based poly(ethylene glycol) hydrogel is applied as a porous scaffold material using indirect solid freeform fabrication (SFF). This approach combines the benefits of SFF with a large freedom in material selection and applicable concentration ranges. A sacrificial 3D poly(ε-caprolactone) structure is generated using fused deposition modeling and used as template to produce hydrogel scaffolds. By changing the template plotting parameters, the scaffold channel sizes vary from 280 to 360 μm, and the strut diameters from 340 to 400 μm. This enables the production of scaffolds with tunable mechanical properties, characterized by an average hardness ranging from 9 to 43 N and from 1 to 6 N for dry and hydrated scaffolds, respectively. Experiments using mouse calvaria preosteoblasts indicate that a gelatin methacrylamide coating of the scaffolds results in an increased cell adhesion and proliferation with improved cell morphology.