Merck
  • Oncogenic tyrosine kinase of malignant hemopathy targets the centrosome.
Oncogenic tyrosine kinase of malignant hemopathy targets the centrosome.
Cancer research (2005-08-17)
Bénédicte Delaval, Sébastien Létard, Hélène Lelièvre, Véronique Chevrier, Laurent Daviet, Patrice Dubreuil, Daniel Birnbaum
ABSTRACT
Myeloproliferative disorders (MPD) are malignant diseases of hematopoietic progenitor cells. Many MPDs result from a chromosomal translocation that creates a fusion gene encoding a chimeric kinase. The fibroblast growth factor receptor 1 (FGFR1)-MPD is characterized by the fusion of the FGFR1 kinase with various partners, including FOP. We show here that both normal FOP and FOP-FGFR1 fusion kinase localize to the centrosome. The fusion kinase encounters substrates at the centrosome where it induces strong phosphorylation on tyrosine residues. Treatment with FGFR1 kinase inhibitor SU5402 abolishes FOP-FGFR1-induced centrosomal phosphorylation and suppresses the proliferative and survival potentials of FOP-FGFR1 Ba/F3 cells. We further show that FOP-FGFR1 allows cells to overcome G1 arrest. Therefore, the FOP-FGFR1 fusion kinase targets the centrosome, activates signaling pathways at this organelle, and sustains continuous entry in the cell cycle. This could represent a potential new mechanism of oncogenic transformation occurring specifically at the centrosome.
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution