Merck
  • Assessing the contribution of thrombospondin-4 induction and ATF6α activation to endoplasmic reticulum expansion and phenotypic modulation in bladder outlet obstruction.
Assessing the contribution of thrombospondin-4 induction and ATF6α activation to endoplasmic reticulum expansion and phenotypic modulation in bladder outlet obstruction.
Scientific reports (2016-09-02)
Katarzyna K Krawczyk, Mari Ekman, Catarina Rippe, Mario Grossi, Bengt-Olof Nilsson, Sebastian Albinsson, Bengt Uvelius, Karl Swärd
ABSTRACT
Phenotypic modulation of smooth muscle cells is a hallmark of disease. The associated expansion of endoplasmic reticulum (ER) volume remains unexplained. Thrombospondin-4 was recently found to promote ATF6α activation leading to ER expansion. Using bladder outlet obstruction as a paradigm for phenotypic modulation, we tested if thrombospondin-4 is induced in association with ATF6α activation and ER expansion. Thrombospondin-4 was induced and ATF6α was activated after outlet obstruction in rodents. Increased abundance of spliced of Xbp1, another ER-stress sensor, and induction of Atf4 and Creb3l2 was also seen. Downstream of ATF6α, Calr, Manf, Sdf2l1 and Pdi increased as did ER size, whereas contractile markers were reduced. Overexpression of ATF6α, but not of thrombospondin-4, increased Calr, Manf, Sdf2l1 and Pdi and caused ER expansion, but the contractile markers were inert. Knockout of thrombospondin-4 neither affected bladder growth nor expression of ATF6α target genes, and repression of contractile markers was the same, even if ATF6α activation was curtailed. Increases of Xbp1s, Atf4 and Creb3l2 were similar. Our findings demonstrate reciprocal regulation of the unfolded protein response, including ATF6α activation and ER expansion, and reduced contractile differentiation in bladder outlet obstruction occurring independently of thrombospondin-4, which however is a sensitive indicator of obstruction.
Sigma-Aldrich
Anti-SDF2L1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Pricing and availability is not currently available.