Merck
  • Ultrastructural relationship of the phagophore with surrounding organelles.
Ultrastructural relationship of the phagophore with surrounding organelles.
Autophagy (2015-02-26)
Joanna Biazik, Päivi Ylä-Anttila, Helena Vihinen, Eija Jokitalo, Eeva-Liisa Eskelinen
ABSTRACT
Phagophore nucleates from a subdomain of the endoplasmic reticulum (ER) termed the omegasome and also makes contact with other organelles such as mitochondria, Golgi complex, plasma membrane and recycling endosomes during its formation. We have used serial block face scanning electron microscopy (SB-EM) and electron tomography (ET) to image phagophore biogenesis in 3 dimensions and to determine the relationship between the phagophore and surrounding organelles at high resolution. ET was performed to confirm whether membrane contact sites (MCSs) are evident between the phagophore and those surrounding organelles. In addition to the known contacts with the ER, we identified MCSs between the phagophore and membranes from putative ER exit sites, late endosomes or lysosomes, the Golgi complex and mitochondria. We also show that one phagophore can have simultaneous MCSs with more than one organelle. Future membrane flux experiments are needed to determine whether membrane contacts also signify lipid translocation.
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Anti-WIPI2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2