- Human promyelocytic leukemia protein is targeted to distinct subnuclear domains in plant nuclei and colocalizes with nucleolar constituents in a SUMO-dependent manner.
Human promyelocytic leukemia protein is targeted to distinct subnuclear domains in plant nuclei and colocalizes with nucleolar constituents in a SUMO-dependent manner.
FEBS open bio (2016-11-12)
Christian E Lamm, Myriam Scherer, Nina Reuter, Bushra Amin, Thomas Stamminger, Uwe Sonnewald
PMID27833854
ABSTRACT
Eukaryotic nuclei are subdivided into subnuclear structures. Among the most prominent of these structures are the nucleolus and the PML nuclear bodies (PML-NBs). PML-NBs are spherical multiprotein aggregates of varying size localized in the interchromosomal area. PML-NB formation is dependent on the presence of the promyelocytic leukemia protein (PML) as well as on post-translational modification of core components by covalent attachment of the small ubiquitin-like modifier (SUMO). So far, PML-NBs as well as PML have been described in mammalian cells only, whereas no orthologs are known in the plant kingdom. In order to investigate conserved mechanisms in PML targeting, we expressed human PML (hPML) fused to the GFP in