Merck
  • Truncated activin type I receptor Alk4 isoforms are dominant negative receptors inhibiting activin signaling.
Truncated activin type I receptor Alk4 isoforms are dominant negative receptors inhibiting activin signaling.
Molecular endocrinology (Baltimore, Md.) (2000-12-16)
Y Zhou, H Sun, D C Danila, S R Johnson, D P Sigai, X Zhang, A Klibanski
ABSTRACT
Activin, a member of the transforming growth factor beta (TGFbeta) superfamily of cytokines, inhibits cell proliferation in a variety of cell types. The functions of activin are mediated by type I and type II serine/threonine kinase receptors. The main type I receptor mediating activin signaling in human cells is ActRIB, also called Alk4. We have previously reported that several truncated Alk4 receptor isoforms are exclusively expressed in human pituitary tumors, and that the majority of such tumors did not exhibit activin-induced growth arrest in culture. We therefore studied the function of these truncated receptor isoforms. Transient expression of these truncated receptors inhibited activin-activated transcription from an activin-responsive reporter construct, 3TPLux. When each of these truncated Alk4 receptors was stably transfected into K562 cells, activin-induced expression of an endogenous gene, junB, was blocked, indicating that inhibition of gene expression also occurred at the chromosomal level. Furthermore, activin administration failed to cause growth inhibition and an increase of the G1 population in these cells. Coimmunoprecipitation experiments showed that the truncated Alk4 receptors formed complexes with type II activin receptors, but were not phosphorylated. These data indicate that the truncated activin type I receptors, predominantly expressed in human pituitary adenomas, function as dominant negative receptors to interfere with wild-type receptor function and block the antiproliferative effect of activin. This may contribute to uncontrolled pituitary cell growth and the development of human pituitary tumors.
Sigma-Aldrich
ALK4 (150-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution