- Ghrelin increases growth hormone production and functional expression of NaV1.1 and Na V1.2 channels in pituitary somatotropes.
Ghrelin increases growth hormone production and functional expression of NaV1.1 and Na V1.2 channels in pituitary somatotropes.
A variety of ion channels are expressed in the plasma membrane of somatotropes within the anterior pituitary gland. Modification of these channels is linked to intracellular Ca2+ levels and therefore to hormone secretion. Previous investigations have shown that the gut-derived orexigenic peptide hormone ghrelin and synthetic GH-releasing peptides (GHRPs) stimulate release of growth hormone (GH) and increase the number of functional voltage-gated Ca2+ and Na+ channels in the membrane of clonal GC somatotropes. Here, we reveal that chronic treatment with ghrelin and its synthetic analog GHRP-6 also increases GH release from bovine pituitary somatotropes in culture, and that this action is associated with a significant increase in Na+ macroscopic current. Consistent with this, Na+ current blockade with tetrodotoxin (TTX) abolished the ghrelin- and GHRP-6-induced increase in GH release. Furthermore, semi-quantitative and real-time RT-PCR analysis revealed an upregulation in the transcript levels of GH, as well as of NaV1.1 and NaV1.2, two isoforms of TTX-sensitive Na+ channels expressed in somatotropes, after treatment with ghrelin or GHRP-6. These findings improve our knowledge on (i) the cellular mechanisms involved in the control of GH secretion, (ii) the molecular diversity of Na+ channels in pituitary somatotropes, and (iii) the regulation of GH and Na+ channel gene expression by ghrelin and GHRPs.