Merck
  • Access to Different Isomeric Dibenzoxazepinones through Copper-Catalyzed C-H Etherification and C-N Bond Construction with Controllable Smiles Rearrangement.
Access to Different Isomeric Dibenzoxazepinones through Copper-Catalyzed C-H Etherification and C-N Bond Construction with Controllable Smiles Rearrangement.
Organic letters (2016-01-16)
Yunfei Zhou, Jianming Zhu, Bo Li, Yong Zhang, Jia Feng, Adrian Hall, Jiye Shi, Weiliang Zhu
ABSTRACT
An efficient new way to access two regio-isomeric dibenzoxazepinones is reported from 8-aminoquinoline benzamides and 2-bromophenols. Through choice of conditions, the reaction proceeds either through a sequential C-H etherification and subsequent Goldberg reaction, both controlled by the aminoquinoline group and Cu(I), or via a C-H etherification and subsequent Smiles rearrangement promoted by Cu(II) and t-BuOK. The 8-aminoquinoline moiety, e.g., 8-amino-5-methoxyquinoline, is readily removable from the structures of dibenzoxazepinones under moderate conditions.
Sigma-Aldrich
8-Amino-5-methoxyquinoline, 95%