- Asymmetric synthesis of new beta,beta-difluorinated cyclic quaternary alpha-amino acid derivatives.
Asymmetric synthesis of new beta,beta-difluorinated cyclic quaternary alpha-amino acid derivatives.
Organic letters (2006-08-25)
Santos Fustero, María Sanchez-Roselló, Vanessa Rodrigo, Carlos del Pozo, Juan F Sanz-Cervera, Antonio Simón, Carmen Ramírez de Arellano
PMID16928091
ABSTRACT
The synthesis of new beta,beta-difluorinated cyclic quaternary alpha-amino acid derivatives 1 in which a ring-closing metathesis reaction (RCM) constitutes the key step is described. The approach employs imidoyl chlorides 3 as fluorinated building blocks, and the overall process involves the stereoselective creation of a quaternary stereocenter. Complete selectivity was achieved when (R)-phenylglycinol methyl ether was used as chiral auxiliary, allowing for the preparation of new six-membered cyclic fluorinated alpha-amino acids as single enantiomers.