Merck
  • Long-term in vitro culture of grape berries and its application to assess the effects of sugar supply on anthocyanin accumulation.
Long-term in vitro culture of grape berries and its application to assess the effects of sugar supply on anthocyanin accumulation.
Journal of experimental botany (2014-01-31)
Zhan Wu Dai, Messaoud Meddar, Christel Renaud, Isabelle Merlin, Ghislaine Hilbert, Serge Delrot, Eric Gomès
ABSTRACT
Grape berry development and ripening are under complex regulation by the nutrients, hormones, and environment cues sensed by the berry. However, the biochemical and molecular mechanisms underlying these types of regulation are poorly understood. A simplified but realistic model system that enables fruit growth conditions to be modulated easily will facilitate the deciphering of these mechanisms. Here, an in vitro culture system of intact detached grape berries was developed by coupling the production of greenhouse fruiting-cuttings and in vitro organ culture techniques. (13)C and (15)N labelling experiments showed that this system enables the intact detached berries actively to absorb and utilize carbon and nitrogen from the culture medium. It was further used to study the effects of sugars on anthocyanin accumulation. A sucrose concentration >2% could induce anthocyanin synthesis in the absence of additional exogenous abscisic acid. The higher the sucrose concentration, the earlier was the induction of anthocyanin accumulation. Glucose, fructose, and sucrose increased anthocyanin accumulation, with glucose and fructose being more effective than sucrose. This increase was not due to an increase in its precursor level, since the phenylalanine content was decreased by a high sugar supply. Instead, genome-wide transcriptome analysis suggests that the sugar-induced enhancement of anthocyanin accumulation results from altered expression of regulatory and structural genes (especially UDP-glucose:anthocyanidin 3-O-glucosyltransferase), together with massive reprogramming in signalling transduction pathways. This in vitro system may serve to study the response of berry composition to nutrient factors and hormones, and their interaction with environmental factors (e.g. light and temperature), which can all be finely tuned and controlled.