Merck
Total synthesis of zincophorin and its methyl ester.
The Journal of organic chemistry (2004-07-03)
Magali Defosseux, Nicolas Blanchard, Christophe Meyer, Janine Cossy
ABSTRACT
A total synthesis of the naturally occurring ionophore zincophorin has been realized. The route features an intramolecular oxymercuration of a cyclopropanemethanol and a Carroll-Claisen rearrangement for the respective elaboration of the C1-C12 and C13-C25 subunits, which have been assembled by using a highly diastereoselective titanium-mediated aldol condensation.
Sigma-Aldrich
Benzyl 3-bromopropyl ether, 98%