Merck
  • P-Selectin Sustains Extramedullary Hematopoiesis in the Gata1 low Model of Myelofibrosis.
P-Selectin Sustains Extramedullary Hematopoiesis in the Gata1 low Model of Myelofibrosis.
Stem cells (Dayton, Ohio) (2015-10-07)
Gerald J Spangrude, Daniel Lewandowski, Fabrizio Martelli, Manuela Marra, Maria Zingariello, Laura Sancillo, Rosa Alba Rana, Anna Rita Migliaccio
ABSTRACT
Splenomegaly is a major manifestation of primary myelofibrosis (PMF) contributing to clinical symptoms and hematologic abnormalities. The spleen from PMF patients contains increased numbers of hematopoietic stem cells (HSC) and megakaryocytes (MK). These MK express high levels of P-selectin (P-sel) that, by triggering neutrophil emperipolesis, may cause TGF-β release and disease progression. This hypothesis was tested by deleting the P-sel gene in the myelofibrosis mouse model carrying the hypomorphic Gata1(low) mutation that induces megakaryocyte abnormalities that recapitulate those observed in PMF. P-sel(null) Gata1(low) mice survived splenectomy and lived 3 months longer than P-sel(WT) Gata1(low) littermates and expressed limited fibrosis and osteosclerosis in the marrow or splenomegaly. Furthermore, deletion of P-sel disrupted megakaryocyte/neutrophil interactions in spleen, reduced TGF-β content, and corrected the HSC distribution that in Gata1(low) mice, as in PMF patients, is abnormally expanded in spleen. Conversely, pharmacological inhibition of TGF-β reduced P-sel expression in MK and corrected HSC distribution. Spleens, but not marrow, of Gata1(low) mice contained numerous cKIT(pos) activated fibrocytes, probably of dendritic cell origin, whose membrane protrusions interacted with MK establishing niches hosting immature cKIT(pos) hematopoietic cells. These activated fibrocytes were not detected in spleens from P-sel(null) Gata1(low) or TGF-β-inhibited Gata1(low) littermates and were observed in spleen, but not in marrow, from PMF patients. Therefore, in Gata1(low) mice, and possibly in PMF, abnormal P-sel expression in MK may mediate the pathological cell interactions that increase TGF-β content in MK and favor establishment of a microenvironment that supports myelofibrosis-related HSC in spleen.
Sigma-Aldrich
Anti-Mouse Collagen Type I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid