Merck
New insights into shear-sensitivity in dinoflagellate microalgae.
Bioresource technology (2015-11-12)
J J Gallardo-Rodríguez, L López-Rosales, A Sánchez-Mirón, F García-Camacho, E Molina-Grima, J J Chalmers
ABSTRACT
A modification of a flow contraction device was used to subject shear-sensitive microalgae to well-defined hydrodynamic forces. The aim of the study was to elucidate if the inhibition of shear-induced growth commonly observed in dinoflagellate microalgae is in effect due to cell fragility that results in cell breakage even at low levels of turbulence. The microalgae assayed did not show any cell breakage even at energy dissipation rates (EDR) around 10(12)Wm(-3), implausible in culture devices. Conversely, animal cells, tested for comparison purposes, showed high physical cell damage at average EDR levels of 10(7)Wm(-3). Besides, very short exposures to high levels of EDR promoted variations in the membrane fluidity of the microalgae assayed, which might trigger mechanosensory cellular mechanisms. Average EDR values of only about 4·10(5)Wm(-3) increased cell membrane fluidity in microalgae whereas, in animal cells, they did not.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture