Merck
  • Potential diagnostic value of regional myocardial adrenergic imaging using (123)I-MIBG SPECT to identify patients with Lewy body diseases.
Potential diagnostic value of regional myocardial adrenergic imaging using (123)I-MIBG SPECT to identify patients with Lewy body diseases.
European journal of nuclear medicine and molecular imaging (2015-01-30)
Adrien Lebasnier, Guillaume Lamotte, Alain Manrique, Damien Peyronnet, Gerard Bouvard, Gilles Defer, Denis Agostini
ABSTRACT
The aim of this study was to determine the potential diagnostic value of regional myocardial adrenergic (123)I-metaiodobenzylguanidine (MIBG) single photon emission computed tomography (SPECT) imaging to identify patients with Lewy body diseases (LBD+). Sixty-four consecutive patients who underwent cardiac (123)I-MIBG SPECT to differentiate LBD+, including Parkinson's disease (PD) and dementia with Lewy bodies (DLB), from patients without LBD (LBD-) were retrospectively reviewed. A neurologist expert in memory disorders determined the final clinical diagnosis by using international clinical diagnostic criteria. Planar [heart to mediastinum ratio (HMR)] and (123)I-MIBG SPECT[(innervation defect score (IDS)] using the 17-segment left ventricular model (five-point scale) were obtained 4 h after the injection of (123)I-MIBG on a low-energy high-resolution (LEHR) collimator. Receiver-operating characteristic (ROC) analysis was performed to determine the optimal HMR and IDS cut-off values to discriminate LBD+ from LBD-. Of the 64 patients, 45 (70 %) were diagnosed LBD+ (DLB, n = 27; PD, n = 18) and 19 were diagnosed LBD- (5 other dementias, 14 other parkinsonisms). The HMR and IDS of LBD+ were significantly different from those of LBD- (1.30 ± 0.21 vs 1.65 ± 0.26, p < 0.001; 39 ± 28 vs 8 ± 16, p = 0.001). The optimal HMR and IDS cut-off values to discriminate LBD+ (n = 45) from LBD- (n = 19) were 1.47 and 6/68, providing a sensitivity and specificity of 82.2 and 84.2% and 86.7 and 73.7%, respectively. Regional myocardial adrenergic (123)I-MIBG imaging SPECT has a potential diagnostic value to identify LBD+.
Sigma-Aldrich
m-Iodobenzylguanidine hemisulfate salt, ≥98% (HPLC and TLC)