Merck
Fabrication and magnetic properties of arrays of metallic nanowires.
Science (New York, N.Y.) (1993-09-03)
T M Whitney, P C Searson, J S Jiang, C L Chien
ABSTRACT
Arrays of ferromagnetic nickel and cobalt nanowires have been fabricated by electrochemical deposition of the metals into templates with nanometer-sized pores prepared by nuclear track etching. These systems display distinctive characteristics because of their one-dimensional microstructure. The preferred magnetization direction is perpendicular to the film plane. Enhanced coercivities as high as 680 oersteds and remnant magnetization up to 90 percent have also been observed.